Learn More
Bioss KD-Validated Adiponectin recep
SDP
Catalog No. 5000064916
Encompass_Preferred
Adiponectin Receptor 1 Primary Antibody Unconjugated Public Immunogen Range 1-46/375 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64916 Supplier Bioss Supplier No. BSM90152R
Adiponectin Receptor 1 Primary Antibody Unconjugated Public Immunogen Range 1-46/375 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide