Learn More
Bioss KD-Validated ADK Rabbit Recomb
SDP
Catalog No. 5000066981
Encompass_Preferred
Adenosine Kinase Primary Antibody Unconjugated Public Immunogen Range 1-85/389 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66981 Supplier Bioss Supplier No. BSM90524R
Adenosine Kinase Primary Antibody Unconjugated Public Immunogen Range 1-85/389 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide