Learn More
Bioss KD-Validated ADP ribosylation
SDP
Catalog No. 5000066797
Encompass_Preferred
ADP Ribosylation Factor 1 Primary Antibody Unconjugated Public Immunogen Range 70-108/181 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66797 Supplier Bioss Supplier No. BSM90071R
ADP Ribosylation Factor 1 Primary Antibody Unconjugated Public Immunogen Range 70-108/181 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide