Learn More
Bioss KD-Validated AMPK alpha 1 Rabb
SDP
Supplier:  Bioss BSM90131R
Encompass_Preferred
Protein Kinase AMP-Activated Catalytic Subunit Alpha 1 Primary Antibody Unconjugated Public Immunogen Range 500-559/559 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65032