Learn More
Bioss KD-Validated Androgen receptor
SDP
Catalog No. 5000065891
Encompass_Preferred
Androgen Receptor Primary Antibody Unconjugated Public Immunogen Range 1-61/920 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65891 Supplier Bioss Supplier No. BSM90059R
Androgen Receptor Primary Antibody Unconjugated Public Immunogen Range 1-61/920 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide