Learn More
Bioss KD-Validated Angio associated
SDP
Catalog No. 5000066989
Encompass_Preferred
Angio Associated Migratory Cell Protein Primary Antibody Unconjugated Public Immunogen Range 15-51/434 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66989 Supplier Bioss Supplier No. BSM90334R
Angio Associated Migratory Cell Protein Primary Antibody Unconjugated Public Immunogen Range 15-51/434 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide