Learn More
ABclonal Technology KD Validated Annexin A1/ANXA1
SDP
Catalog No. 5000012639
Encompass_Preferred
This gene encodes a membrane-localized protein that binds phospholipids This protein inhibits phospholipase A2 and has anti-inflammatory activity Loss of function or expression of this gene has been detected in multiple tumors
Catalog No. 50-000-12639 Supplier ABclonal Technology Supplier No. A25918100UL
This gene encodes a membrane-localized protein that binds phospholipids This protein inhibits phospholipase A2 and has anti-inflammatory activity Loss of function or expression of this gene has been detected in multiple tumors