Learn More
Bioss KD-Validated Annexin A10 Rabbi
SDP
Catalog No. 5000066915
Encompass_Preferred
Annexin A10 Primary Antibody Unconjugated Public Immunogen Range 50-200/324 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66915 Supplier Bioss Supplier No. BSM90154R
Annexin A10 Primary Antibody Unconjugated Public Immunogen Range 50-200/324 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide