Learn More
Bioss KD-Validated Annexin V Rabbit
SDP
Catalog No. 5000066616
Encompass_Preferred
Annexin A5 Primary Antibody Unconjugated Public Immunogen Range 301-320/320 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66616 Supplier Bioss Supplier No. BSM90353R
Annexin A5 Primary Antibody Unconjugated Public Immunogen Range 301-320/320 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide