Learn More
Bioss KD-Validated AP2S1 Rabbit Reco
SDP
Catalog No. 5000065012
Encompass_Preferred
Adaptor Related Protein Complex 2 Subunit Sigma 1 Primary Antibody Unconjugated Public Immunogen Range 110-142/142 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65012 Supplier Bioss Supplier No. BSM90048R
Adaptor Related Protein Complex 2 Subunit Sigma 1 Primary Antibody Unconjugated Public Immunogen Range 110-142/142 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide