Learn More
Bioss KD-Validated APBB1 Rabbit Reco
SDP
Catalog No. 5000067000
Encompass_Preferred
Amyloid Beta Precursor Protein Binding Family B Member 1 Primary Antibody Unconjugated Public Immunogen Range 650-710/710 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67000 Supplier Bioss Supplier No. BSM90097R
Amyloid Beta Precursor Protein Binding Family B Member 1 Primary Antibody Unconjugated Public Immunogen Range 650-710/710 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide