Learn More
Bioss KD-Validated APPL1 Rabbit Reco
SDP
Catalog No. 5000065915
Encompass_Preferred
DCC-interacting protein 13-alpha Primary Antibody Unconjugated Public Immunogen Range 400-450/709 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65915 Supplier Bioss Supplier No. BSM90171R
DCC-interacting protein 13-alpha Primary Antibody Unconjugated Public Immunogen Range 400-450/709 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide