Learn More
ABclonal Technology KD Validated Argonaute-2
SDP
Catalog No. 5000012519
Encompass_Preferred
This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference The encoded protein is highly basic and contains a PAZ domain and a PIWI domain It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12519 Supplier ABclonal Technology Supplier No. A25642100UL
This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference The encoded protein is highly basic and contains a PAZ domain and a PIWI domain It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing Multiple transcript variants encoding different isoforms have been found for this gene