Learn More
Bioss KD-Validated Argonaute RISC ca
SDP
Catalog No. 5000066595
Encompass_Preferred
Argonaute RISC Catalytic Component 2 Primary Antibody Unconjugated Public Immunogen Range 370-420/859 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66595 Supplier Bioss Supplier No. BSM90315R
Argonaute RISC Catalytic Component 2 Primary Antibody Unconjugated Public Immunogen Range 370-420/859 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide