Learn More
Bioss KD-Validated ARPC2 Rabbit Reco
SDP
Catalog No. 5000064911
Encompass_Preferred
Actin Related Protein 2/3 Complex Subunit 2 Primary Antibody Unconjugated Public Immunogen Range 230-300/300 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64911 Supplier Bioss Supplier No. BSM90150R
Actin Related Protein 2/3 Complex Subunit 2 Primary Antibody Unconjugated Public Immunogen Range 230-300/300 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide