Learn More
Bioss KD-Validated Arrestin beta 1 R
SDP
Catalog No. 5000065437
Encompass_Preferred
Arrestin Beta 1 Primary Antibody Unconjugated Public Immunogen Range 1-48/418 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65437 Supplier Bioss Supplier No. BSM90361R
Arrestin Beta 1 Primary Antibody Unconjugated Public Immunogen Range 1-48/418 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide