Learn More
Bioss KD-Validated Aryl hydrocarbon
SDP
Catalog No. 5000065025
Encompass_Preferred
Aryl Hydrocarbon Receptor Primary Antibody Unconjugated Public Immunogen Range 724-780/848 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65025 Supplier Bioss Supplier No. BSM90064R
Aryl Hydrocarbon Receptor Primary Antibody Unconjugated Public Immunogen Range 724-780/848 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide