Learn More
Bioss KD-Validated Atg4C Rabbit Reco
SDP
Catalog No. 5000066892
Encompass_Preferred
Autophagy Related 4C Cysteine Peptidase Primary Antibody Unconjugated Public Immunogen Range 350-458/458 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66892 Supplier Bioss Supplier No. BSM90186R
Autophagy Related 4C Cysteine Peptidase Primary Antibody Unconjugated Public Immunogen Range 350-458/458 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide