Learn More
Bioss KD-Validated ATG5 Rabbit Recom
SDP
Catalog No. 5000064931
Encompass_Preferred
Autophagy Related 5 Primary Antibody Unconjugated Public Immunogen Range 1-64/275 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64931 Supplier Bioss Supplier No. BSM90185R
Autophagy Related 5 Primary Antibody Unconjugated Public Immunogen Range 1-64/275 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide