Learn More
Bioss KD-Validated ATG9A Rabbit Reco
SDP
Catalog No. 5000065889
Encompass_Preferred
Autophagy Related 9A Primary Antibody Unconjugated Public Immunogen Range 812-839/839 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65889 Supplier Bioss Supplier No. BSM90069R
Autophagy Related 9A Primary Antibody Unconjugated Public Immunogen Range 812-839/839 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide