Learn More
Bioss KD-Validated ATP citrate lyase
SDP
Catalog No. 5000064983
Encompass_Preferred
ATP Citrate Lyase Primary Antibody Unconjugated Public Immunogen Range 1050-1101/1101 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64983 Supplier Bioss Supplier No. BSM90319R
ATP Citrate Lyase Primary Antibody Unconjugated Public Immunogen Range 1050-1101/1101 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide