Learn More
Bioss KD-Validated ATP1B3 Rabbit Rec
SDP
Catalog No. 5000065417
Encompass_Preferred
ATPase Na /K Transporting Subunit Beta 3 Primary Antibody Unconjugated Public Immunogen Range 241-279/279 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65417 Supplier Bioss Supplier No. BSM90208R
ATPase Na /K Transporting Subunit Beta 3 Primary Antibody Unconjugated Public Immunogen Range 241-279/279 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide