Learn More
Bioss KD-Validated ATP5B Rabbit Reco
SDP
Catalog No. 5000064913
Encompass_Preferred
ATP Synthase F1 Subunit Beta Primary Antibody Unconjugated Public Immunogen Range 140-200/529 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64913 Supplier Bioss Supplier No. BSM90158R
ATP Synthase F1 Subunit Beta Primary Antibody Unconjugated Public Immunogen Range 140-200/529 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide