Learn More
Bioss KD-Validated ATP7B Rabbit Reco
SDP
Catalog No. 5000065184
Encompass_Preferred
ATPase Copper Transporting Beta Primary Antibody Unconjugated Public Immunogen Range 1400-1465/1465 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65184 Supplier Bioss Supplier No. BSM90170R
ATPase Copper Transporting Beta Primary Antibody Unconjugated Public Immunogen Range 1400-1465/1465 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide