Learn More
Bioss KD-Validated BCAR1 Rabbit Reco
SDP
Catalog No. 5000065125
Encompass_Preferred
Breast cancer anti-estrogen resistance protein 1 Primary Antibody Unconjugated Public Immunogen Range 1-68/870 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65125 Supplier Bioss Supplier No. BSM90095R
Breast cancer anti-estrogen resistance protein 1 Primary Antibody Unconjugated Public Immunogen Range 1-68/870 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide