Learn More
Bioss KD-Validated Bone morphogeneti
SDP
Catalog No. 5000066209
Encompass_Preferred
Bone Morphogenetic Protein 6 Primary Antibody Unconjugated Public Immunogen Range 330-430/513 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66209 Supplier Bioss Supplier No. BSM90328R
Bone Morphogenetic Protein 6 Primary Antibody Unconjugated Public Immunogen Range 330-430/513 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide