Learn More
Bioss KD-Validated Brain derived neu
SDP
Catalog No. 5000066615
Encompass_Preferred
Brain Derived Neurotrophic Factor Primary Antibody Unconjugated Public Immunogen Range 217-230/247 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66615 Supplier Bioss Supplier No. BSM90346R
Brain Derived Neurotrophic Factor Primary Antibody Unconjugated Public Immunogen Range 217-230/247 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide