Learn More
Bioss KD-Validated C1QBP Rabbit Reco
SDP
Catalog No. 5000066918
Encompass_Preferred
Complement C1q Binding Protein Primary Antibody Unconjugated Public Immunogen Range 200-250/282 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66918 Supplier Bioss Supplier No. BSM90164R
Complement C1q Binding Protein Primary Antibody Unconjugated Public Immunogen Range 200-250/282 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide