Learn More
Bioss KD-Validated C4BPB Rabbit Reco
SDP
Catalog No. 5000066894
Encompass_Preferred
Complement Component 4 Binding Protein Beta Primary Antibody Unconjugated Public Immunogen Range 80-128/128 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66894 Supplier Bioss Supplier No. BSM90392R
Complement Component 4 Binding Protein Beta Primary Antibody Unconjugated Public Immunogen Range 80-128/128 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide