Learn More
Bioss KD-Validated Calpain 1 Rabbit
SDP
Catalog No. 5000065005
Encompass_Preferred
Calpain 1 Primary Antibody Unconjugated Public Immunogen Range 154-200/714 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65005 Supplier Bioss Supplier No. BSM90077R
Calpain 1 Primary Antibody Unconjugated Public Immunogen Range 154-200/714 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide