Learn More
Bioss KD-Validated Calpain 2 Rabbit
SDP
Catalog No. 5000066794
Encompass_Preferred
Calpain 2 Primary Antibody Unconjugated Public Immunogen Range 657-700/700 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66794 Supplier Bioss Supplier No. BSM90079R
Calpain 2 Primary Antibody Unconjugated Public Immunogen Range 657-700/700 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide