Learn More
Bioss KD-Validated Calpain small sub
SDP
Catalog No. 5000065122
Encompass_Preferred
Calpain Small Subunit 1 Primary Antibody Unconjugated Public Immunogen Range 50-100/268 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65122 Supplier Bioss Supplier No. BSM90103R
Calpain Small Subunit 1 Primary Antibody Unconjugated Public Immunogen Range 50-100/268 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide