Learn More
Bioss KD-Validated Calreticulin Rabb
SDP
Catalog No. 5000065888
Encompass_Preferred
Calreticulin Primary Antibody Unconjugated Public Immunogen Range 45-86/417 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65888 Supplier Bioss Supplier No. BSM90012R
Calreticulin Primary Antibody Unconjugated Public Immunogen Range 45-86/417 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide