Learn More
Bioss KD-Validated Carbamoyl-phospha
SDP
Catalog No. 5000065165
Encompass_Preferred
Carbamoyl-Phosphate Synthase 1 Primary Antibody Unconjugated Public Immunogen Range 100-166/1500 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65165 Supplier Bioss Supplier No. BSM90036R
Carbamoyl-Phosphate Synthase 1 Primary Antibody Unconjugated Public Immunogen Range 100-166/1500 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide