Learn More
Bioss KD-Validated Carbonic anhydras
SDP
Catalog No. 5000066637
Encompass_Preferred
Carbonic Anhydrase 2 Primary Antibody Unconjugated Public Immunogen Range 574-599/599 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66637 Supplier Bioss Supplier No. BSM90401R
Carbonic Anhydrase 2 Primary Antibody Unconjugated Public Immunogen Range 574-599/599 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide