Learn More
Bioss KD-Validated Carbonic anhydras
SDP
Catalog No. 5000066531
Encompass_Preferred
Carbonic Anhydrase 9 Primary Antibody Unconjugated Public Immunogen Range 204-256/459 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66531 Supplier Bioss Supplier No. BSM90340R
Carbonic Anhydrase 9 Primary Antibody Unconjugated Public Immunogen Range 204-256/459 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide