Learn More
Bioss KD-Validated Caspase 2 Rabbit
SDP
Catalog No. 5000066896
Encompass_Preferred
Caspase 2 Primary Antibody Unconjugated Public Immunogen Range 84-98/331 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66896 Supplier Bioss Supplier No. BSM90411R
Caspase 2 Primary Antibody Unconjugated Public Immunogen Range 84-98/331 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide