Learn More
Bioss KD-Validated Caspase 8 Rabbit
SDP
Catalog No. 5000066612
Encompass_Preferred
Caspase 8 Primary Antibody Unconjugated Public Immunogen Range 200-250/479 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66612 Supplier Bioss Supplier No. BSM90337R
Caspase 8 Primary Antibody Unconjugated Public Immunogen Range 200-250/479 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide