Learn More
Bioss KD-Validated Catalase Rabbit R
SDP
Catalog No. 5000065011
Encompass_Preferred
Catalase Primary Antibody Unconjugated Public Immunogen Range 50-100/527 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65011 Supplier Bioss Supplier No. BSM90045R
Catalase Primary Antibody Unconjugated Public Immunogen Range 50-100/527 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide