Learn More
Bioss KD-Validated Catechol-O-methyl
SDP
Catalog No. 5000066804
Encompass_Preferred
Catechol-O-Methyltransferase Primary Antibody Unconjugated Public Immunogen Range 31-87/271 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66804 Supplier Bioss Supplier No. BSM90100R
Catechol-O-Methyltransferase Primary Antibody Unconjugated Public Immunogen Range 31-87/271 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide