Learn More
Bioss KD-Validated CD163 Rabbit Reco
SDP
Catalog No. 5000066857
Encompass_Preferred
CD163 Primary Antibody Unconjugated Public Immunogen Range 1001-1156/1156 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66857 Supplier Bioss Supplier No. BSM90324R
CD163 Primary Antibody Unconjugated Public Immunogen Range 1001-1156/1156 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide