Learn More
Bioss KD-Validated CD166 Rabbit Reco
SDP
Catalog No. 5000067002
Encompass_Preferred
Activated Leukocyte Cell Adhesion Molecule Primary Antibody Unconjugated Public Immunogen Range 540-583/583 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67002 Supplier Bioss Supplier No. BSM90082R
Activated Leukocyte Cell Adhesion Molecule Primary Antibody Unconjugated Public Immunogen Range 540-583/583 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide