Learn More
Bioss KD-Validated CD58 Rabbit Recom
SDP
Catalog No. 5000064876
Encompass_Preferred
CD58 Molecule Primary Antibody Unconjugated Public Immunogen Range 51-82/250 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64876 Supplier Bioss Supplier No. BSM90051R
CD58 Molecule Primary Antibody Unconjugated Public Immunogen Range 51-82/250 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide