Learn More
Bioss KD-Validated CDC16 Rabbit Reco
SDP
Catalog No. 5000066997
Encompass_Preferred
Cell Division Cycle 16 Primary Antibody Unconjugated Public Immunogen Range 200-260/260 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66997 Supplier Bioss Supplier No. BSM90400R
Cell Division Cycle 16 Primary Antibody Unconjugated Public Immunogen Range 200-260/260 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide