Learn More
Bioss KD-Validated CDC25C Rabbit Rec
SDP
Catalog No. 5000065189
Encompass_Preferred
Cell Division Cycle 25C Primary Antibody Unconjugated Public Immunogen Range 11-63/473 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65189 Supplier Bioss Supplier No. BSM90073R
Cell Division Cycle 25C Primary Antibody Unconjugated Public Immunogen Range 11-63/473 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide