Learn More
Bioss KD-Validated CDC73 Rabbit Reco
SDP
Catalog No. 5000064873
Encompass_Preferred
Cell Division Cycle 73 Primary Antibody Unconjugated Public Immunogen Range 350-400/531 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64873 Supplier Bioss Supplier No. BSM90087R
Cell Division Cycle 73 Primary Antibody Unconjugated Public Immunogen Range 350-400/531 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide