Learn More
Bioss KD-Validated Chloride intracel
SDP
Catalog No. 5000067001
Encompass_Preferred
Chloride Intracellular Channel 4 Primary Antibody Unconjugated Public Immunogen Range 1-28/253 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67001 Supplier Bioss Supplier No. BSM90085R
Chloride Intracellular Channel 4 Primary Antibody Unconjugated Public Immunogen Range 1-28/253 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide