Learn More
Bioss KD-Validated Chondroitin sulfa
SDP
Catalog No. 5000065913
Encompass_Preferred
Chondroitin Sulfate Proteoglycan 4 Primary Antibody Unconjugated Public Immunogen Range 2250-2322/2322 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65913 Supplier Bioss Supplier No. BSM90159R
Chondroitin Sulfate Proteoglycan 4 Primary Antibody Unconjugated Public Immunogen Range 2250-2322/2322 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide