Learn More
Bioss KD-Validated Chromodomain heli
SDP
Catalog No. 5000066207
Encompass_Preferred
Chromodomain Helicase DNA Binding Protein 3 Primary Antibody Unconjugated Public Immunogen Range 1940-2000/2000 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66207 Supplier Bioss Supplier No. BSM90314R
Chromodomain Helicase DNA Binding Protein 3 Primary Antibody Unconjugated Public Immunogen Range 1940-2000/2000 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide