Learn More
Bioss KD-Validated Chromodomain heli
SDP
Supplier:  Bioss BSM90046R
Encompass_Preferred
Chromodomain Helicase DNA Binding Protein 4 Primary Antibody Unconjugated Public Immunogen Range 1800-1870/1912 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66845